Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha V beta 3 Antibody (23C6), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002677G
Description
Integrin alpha V beta 3 Monoclonal specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Integrin alpha V beta 3 | |
Monoclonal | |
DyLight 488 | |
antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Reacts with alpha V beta 3. 23C6 may be useful for bone resorption modulation, osteoclast identification, receptor purification, malignant melanoma identification and treatment. Human. Cross-reacts with rabbit and chicken, as reported in scientific literature. | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
23C6 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
ITGAV | |
A cell suspension containing osteoclasts from osteoclastomas. | |
0.1 mL | |
Adaptive Immunity, Angiogenesis, Cancer, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction | |
3685 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction