Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha V beta 3 Antibody (27.1 (VNR-1)) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $792.00
Specifications
Antigen | Integrin alpha V beta 3 |
---|---|
Clone | 27.1 (VNR-1) |
Concentration | 1 mg/mL |
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:1000, Immunohistochemistry-Paraffin (Negative) |
Applications | Immunocytochemistry, Immunoprecipitation, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence |
Description
Integrin alpha V beta 3 Monoclonal specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin (Negative).Specifications
Integrin alpha V beta 3 | |
1 mg/mL | |
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Angiogenesis, Cancer, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction | |
P06756 | |
3685 | |
This antibody is specific to Integrin Alpha VBeta 3 | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
27.1 (VNR-1) | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:1000, Immunohistochemistry-Paraffin (Negative) | |
Monoclonal | |
Purified | |
RUO | |
Human | |
antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 | |
ITGAV | |
IgG1 | |
Protein A purified | |
Reacts with the human integrin alphaVbeta3. Shown to be specific for alphaVbeta3 by Loftus JC et al (1990) Science 249:915-918. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title