Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha V beta 5 Antibody (P5H9), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2528E
Description
Integrin alpha V beta 5 Monoclonal antibody specifically detects Integrin alpha V beta 5 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry, CyTOFSpecifications
Integrin alpha V beta 5 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3685 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry, CyTOF | |
P5H9 | |
Flow Cytometry, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry, CyTOF-ready | |
CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR | |
HT1080 human fibrosarcoma cell line | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction