Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha V beta 6 Antibody (7k), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP250448AF700
Description
Integrin alpha V beta 6 Monoclonal specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.Specifications
Integrin alpha V + beta 6 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
3T3b6, mouse fibroblast expressing human Integrin alpha V beta 6 | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
Does not bind mouse. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
7k | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction