Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha V beta 6 Antibody (Av beta6 62ow.17), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP250450B
Description
Integrin alpha V beta 6 Monoclonal specifically detects Integrin alpha V beta 6 in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Integrin alpha V + beta 6 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
Mouse fibroblasts retrovirally infected with cDNA to human beta6 (3T3 beta6.19). Cells expressed a mouse/human av beta6 chimera. | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
Does not recognize mouse av beta 6. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry | |
Av beta6 62ow.17 | |
Western Blot, Immunohistochemistry | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction