Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha V beta 6 Antibody (Av beta6 62ow.17) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$196.10
Specifications
Antigen | Integrin alpha V beta 6 |
---|---|
Clone | Av beta6 62ow.17 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
Applications | Western Blot, Immunohistochemistry |
Description
Integrin alpha V beta 6 Monoclonal antibody specifically detects Integrin alpha V beta 6 in Human, Mouse (Negative) samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Integrin alpha V beta 6 | |
1 mg/mL | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rat | |
Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
PBS | |
3685 | |
IgG1 | |
Protein G purified |
Av beta6 62ow.17 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse (Negative) | |
CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR | |
Mouse fibroblasts retrovirally infected with cDNA to human beta6 (3T3 beta6.19). Cells expressed a mouse/human av beta6 chimera. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title