Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha V beta 6 Antibody (AvB6 53a.2), Allophycocyanin/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP250449APCCY7
Description
Integrin alpha V beta 6 Monoclonal specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Flow Cytometry.Specifications
Integrin alpha V beta 6 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
ITGAV | |
Mouse fibroblasts retrovirally infected with cDNA to human Integrin alpha V beta 6 (3T3b6.19). Cells expressed a mouse/human Integrin alpha V beta 6 chimera. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Flow Cytometry | |
AvB6 53a.2 | |
Flow Cytometry | |
CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR | |
Rat | |
Protein G purified | |
Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
3685 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction