Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha V/CD51 Antibody (P2W7), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002618PCP
Description
Integrin alpha V/CD51 Monoclonal specifically detects Integrin alpha V/CD51 in Human samples. It is validated for Flow Cytometry.Specifications
Integrin alpha V/CD51 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
ITGAV | |
An ocular melanoma cell line (V+B2) expressing high levels of human integrin alpha V beta 2. | |
0.1 mL | |
Adaptive Immunity, Angiogenesis, Cancer, Cytoskeleton Markers, Immunology, Signal Transduction | |
3685 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
P2W7 | |
Flow Cytometry | |
antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction