Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin B8 Mouse anti-Human, Clone: 11D4, MilliporeSigma™

Catalog No. MABT912MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT912MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT912MI Supplier MilliporeSigma Supplier No. MABT912

Mouse Monoclonal Antibody

This mouse monoclonal Anti-Integrin B8 Antibody, clone11D4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin B8
Applications Western Blot
Classification Monoclonal
Clone 11D4
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ITGB8
Host Species Mouse
Immunogen A truncated recombinant human Integrin B8, isoform 2, corresponding to amino acids 136-769.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_002205
Test Specificity Clone11D4 specifically detects isoform 2 of human integrin beta-8.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.