Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 1/CD29 Antibody (8E3), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250139AF488
Description
Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Integrin beta 1/CD29 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ITGB1 | |
Purified Integrin beta 1/CD29 from HT-1080 cells. | |
0.1 mL | |
Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
3688 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
8000 | |
Western Blot, ELISA, Immunohistochemistry | |
CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Under native conditions, clone 8E3 is shown to recognize the unbent, monomeric ER form of Integrin beta 1/CD29 at the PSI domain; however, the epitope only becomes exposed in the Golgi form after denaturation. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction