Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 1/CD29 Antibody (ITGB1/8744R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32056720UG
Description
Integrin beta 1/CD29 Monoclonal antibody specifically detects Integrin beta 1/CD29 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Integrin beta 1/CD29 | |
Monoclonal | |
0.2 mg/ml | |
ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL | |
CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
Recombinant full-length human Integrin beta 1/CD29 protein | |
20 μg | |
Cancer, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers | |
3688 | |
Store at 4&drg;C. Do not freeze. | |
IgG κ |
ELISA, Immunohistochemistry (Paraffin) | |
ITGB1/8744R | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction