Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 1/CD29 Antibody (JB1B) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Specifications

Antigen Integrin beta 1/CD29
Clone JB1B
Concentration 0.5 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 0.1 ug/ml-1 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1-5 ug, Immunohistochemistry-Frozen 5 ug/ml
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10064804
SDP
View Documents
Novus Biologicals
NB10064804
0.1 mg N/A N/A N/A
Description

Description

Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Human, Porcine, Primate, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications

Specifications

Integrin beta 1/CD29
0.5 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Unconjugated
Mouse
Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Signal Transduction, Stem Cell Markers
P05556
3688
Purified Human beta1 integrin
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
JB1B
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 0.1 ug/ml-1 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1-5 ug, Immunohistochemistry-Frozen 5 ug/ml
Monoclonal
Purified
RUO
Human, Pig, Primate, Rat (Negative)
CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
ITGB1
IgG2a
Protein A or G purified
NB100-64804 reacts with Human Integrin beta1 as confirmed with beta1 transfectants and by reciprocal preclearing of beta1 reactivity with anti-beta1. The antibody stimulates the adherence of several cell types. The removal of azide is recommended prior to use in functional assays.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.