Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 1/CD29 Antibody (P5D2), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB17781MFV500
Description
Integrin beta 1/CD29 Monoclonal antibody specifically detects Integrin beta 1/CD29 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Blocking Assay, Immunocytochemistry, CyTOF, KnockDownSpecifications
| Integrin beta 1/CD29 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 3688 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunoprecipitation, Blocking Assay, Immunocytochemistry, CyTOF | |
| P5D2 | |
| Flow Cytometry, Immunoprecipitation, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready, Knockout Validated | |
| CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
| Human skin keratinocytes | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction