Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Integrin beta-1 (CD29), Mouse, Unlabeled, Clone: 13C4.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABT529
Description
Integrin beta-1 (UniProt P05556; also known as CD29, Glycoprotein Iia, GPIIA, Integrin VLA-4 subunit beta, Very late activation protein, beta polypeptide) is encoded by the ITGB1 (also known as FNRB, MDF2, MSK12, VLA-BETA, VLAB) gene (Gene ID 3688) in human. Integrin beta-1 is a 130kDa transmembrane glycoprotein that interact with various integrin alpha subunits (including alpha 1, alpha 2, alpha 3, alpha 4, alpha 5, and alpha 6) to form the functional receptor complexes that bind to specific extracellular matrix proteins. Integrin receptors regulate a variety of important biological functions, including embryonic development, wound repair, hemostasis, and prevention of programmed cell death. They are also implicated in abnormal pathological states such as tumor directed angiogenesis, tumor growth and metastasis. These heterodimeric receptors bridge the cytoplasmic actin cytoskeleton with proteins present in the extracellular matrix and/or on adjacent cells. Interactions between integrins and the extracellular matrix lead to the activation of signal transduction pathways and regulation of gene expression.
Specifications
Integrin beta-1 (CD29) | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide. | |
P05556 | |
GST-tagged recombinant human Integrin beta-1 fragment corresponding to the membrane-proximal extracellular region. | |
100 μL | |
Cell Structure | |
NP_002202 | |
Human | |
Purified |
Western Blot | |
13C4.1 | |
Unconjugated | |
ITGB1, FNRB, MDF2, MSK12, VLA-BETA, VLAB | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 13C4.1 targets an epitope within the membrane-proximal extracellular region present in all five spliced isoforms of human Integrin beta-1 reported by UniPort (P05556). | |
Stable for 1 year at 2°-8°C from date of receipt. | |
IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction