Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta-1 Mouse anti-Human, Clone: 1A7C9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Integrin beta-1 |
---|---|
Clone | 1A7C9 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6747
![]() ![]() |
Proteintech Group Inc
663151IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Integrins are heterodimeric proteins made up of alpha and beta subunits. At least 18 alpha and 8 beta subunits have been described in mammals. Integrin family members are membrane receptors involved in cell adhesion and recognition in a variety of processes including embryogenesis, hemostasis, tissue repair, immune response and metastatic diffusion of tumor cells. This gene encodes a beta subunit. Multiple alternatively spliced transcript variants which encode different protein isoforms have been found for this gene.Specifications
Integrin beta-1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD29, FNRB, GPIIA, Integrin beta 1, Integrin beta-1, ITGB1, MDF2, MSK12, VLA 4 subunit beta, VLA BETA, VLAB | |
ITGB1 | |
IgG1 | |
Protein G | |
Antibody |
1A7C9 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P05556 | |
3688 | |
Integrin beta-1 Fusion Protein Ag24476 | |
Primary | |
-20°C | |
ITGB1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title