Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 2/CD18 Antibody (M18/2), Janelia Fluor™ 646, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP141272JF646
Description
Integrin beta 2/CD18 Monoclonal specifically detects Integrin beta 2/CD18 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Integrinbeta2/CD18 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ITGB2 | |
Murine cytotoxic T cell glycoproteins | |
0.1 mL | |
Primary | |
The rat monoclonal antibody M18/2 recognizes CD18 antigen (integrin beta2 subunit; beta2 integrin), a 95 kDa type I transmembrane protein expressed on all leukocytes. | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
M18/2 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
Rat | |
Protein G purified | |
Apoptosis, Cytokine Research, Immunology | |
3689 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction