Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 2/CD18 Antibody (YFC118.3) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$207.00 - $501.50
Specifications
Antigen | Integrin beta 2/CD18 |
---|---|
Clone | YFC118.3 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:1000-1:5000 |
Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Flow Cytometry, Immunoprecipitation, Immunocytochemistry |
Description
Integrin beta 2/CD18 Monoclonal specifically detects Integrin beta 2/CD18 in Human, Canine, Guinea Pig samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
Integrin beta 2/CD18 | |
1 mg/mL | |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rat | |
Apoptosis, Cytokine Research, Immunology | |
959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
ITGB2 | |
IgG2b | |
Protein G purified | |
This recognises the human CD18 cell surface antigen, the integrin beta2 subunit. CD18 is expressed by the majority of leucocytes. |
YFC118.3 | |
Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:1000-1:5000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Canine, Guinea Pig | |
3689 | |
Human neutrophils | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title