Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 2/CD18 Mouse anti-Human, Clone: MEM48, Novus Biologicals™

Mouse Monoclonal Antibody
$544.67 - $1113.62
Specifications
| Antigen | Integrin beta 2/CD18 |
|---|---|
| Clone | MEM48 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay |
Description
Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional AssaySpecifications
| Integrin beta 2/CD18 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cytokine Research, Immunology | |
| PBS. Sterile-filtered through 0.22 um. | |
| 3689 | |
| IgG1 κ | |
| Affinity purified | |
| Antibody |
| MEM48 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
| Leucocytes from an LGL - type leukaemia | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title