Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 2/CD18 Rabbit anti-Human, Clone: 60.3, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | Integrin beta 2/CD18 |
---|---|
Clone | 60.3 |
Concentration | 1 mg/ml |
Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Functional (Inhibition), Block/Neutralize |
Applications | Western Blot, ELISA, Immunofluorescence, Inhibition Assays, Neutralization |
Description
Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Inhibition Assays, NeutralizationSpecifications
Integrin beta 2/CD18 | |
1 mg/ml | |
Western Blot, ELISA, Immunofluorescence, Inhibition Assays, Neutralization | |
Unconjugated | |
Rabbit | |
Apoptosis, Cytokine Research, Immunology, Phospho Specific | |
PBS | |
3689 | |
IgG κ | |
Protein A purified |
60.3 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Functional (Inhibition), Block/Neutralize | |
Monoclonal | |
Purified | |
RUO | |
Human | |
959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
The original antibody was generated by immunizing BALB/c mice with alloantigen activated T cells. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title