Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 3/CD61 Antibody (909114), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB41182B
Description
Integrin beta 3/CD61 Monoclonal antibody specifically detects Integrin beta 3/CD61 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
Integrin beta 3/CD61 | |
Monoclonal | |
Biotin | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
909114 | |
Flow Cytometry, CyTOF-ready | |
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 2b and Integrin beta 3 linked heterodimer, Leu32-Arg988 (Integrin alpha 2b) & Glu26-Asp717 (Integrin beta 3), Accession # NP_034705 (Integrin alpha 2b) & O54890 (Integrin beta 3) | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
3690 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction