Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 3/CD61 Antibody (F11), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065387PE
Description
Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Rat samples. It is validated for Flow Cytometry.Specifications
Integrin beta 3/CD61 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
ITGB3 | |
Bone cell suspension from SD rats | |
0.25 mL | |
Cancer, Cytoskeleton Markers, Phospho Specific, Signal Transduction | |
3690 | |
Rat | |
Purified |
Flow Cytometry | |
F11 | |
Flow Cytometry | |
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
NB100-65387 recognises the rat CD61 cell surface antigen, also known as the integrin b3 chain. CD61 is expressed by platelets, megakaryocytes and osteoclasts. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction