Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 3/CD61 Antibody (ITGB3/1713) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | Integrin beta 3/CD61 |
---|---|
Clone | ITGB3/1713 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254562100
![]() |
Novus Biologicals
NBP254562100UG |
100 μg |
Each for $646.00
|
|
|||||
NB254562200
![]() |
Novus Biologicals
NBP254562200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
Integrin beta 3/CD61 | |
1 mg/mL | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Cancer, Cytoskeleton Markers, Signal Transduction | |
PBS with No Preservative | |
3690 | |
Recombinant human ITGB3 protein | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
ITGB3/1713 | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa | |
ITGB3 | |
IgG1 κ | |
Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title