Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 3/CD61 Antibody (ITGB3/1713), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$246.61 - $554.73

Specifications

Antigen Integrin beta 3/CD61
Clone ITGB3/1713
Concentration 0.2 mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25339320
SDP
View Documents
Novus Biologicals
NBP25339320UG
20 μg
Each for $246.61
Only null left
Add to Cart
 
NB253393100
SDP
View Documents
Novus Biologicals
NBP253393100UG
100 μg
Each for $554.73
Only null left
Add to Cart
 
NB253393200
SDP
View Documents
Novus Biologicals
NBP253393200UG
200 μg N/A N/A N/A
Description

Description

Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Integrin beta 3/CD61
0.2 mg/mL
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Cytoskeleton Markers, Phospho Specific, Signal Transduction
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
3690
Recombinant human ITGB3 protein
Primary
Store at 4C.
ITGB3/1713
Flow Cytometry 0.5 - 1 ug/million cells, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa
ITGB3
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.