Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
integrin beta 4 binding protein Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP152641IR
Description
integrin beta 4 binding protein Polyclonal antibody specifically detects integrin beta 4 binding protein in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
integrin beta 4 binding protein | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
b(2)gcn, B(2)GCN homolog, B4 integrin interactor, CAB, EIF3Ap27(BBP), eIF-6, eukaryotic translation initiation factor 3A, eukaryotic translation initiation factor 6, ITGB4BPintegrin beta 4 binding protein, p27 beta-4 integrin-binding protein, p27BBP | |
The immunogen for this product maps to a region between residue 150 and 200 of human Eukaryotic Translation Initiation Factor 6 using the numbering given in entry NP_852133.1 (GeneID 3692). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
3692 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction