Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 4/CD104 Antibody (308601), DyLight 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4054E
Description
Integrin beta 4/CD104 Monoclonal antibody specifically detects Integrin beta 4/CD104 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
Integrin beta 4/CD104 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
308601 | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 4/CD104, Asn29-Ser711, Accession # NP_001005608 | |
0.1 mL | |
Adaptive Immunity, Immunology, Signal Transduction | |
3691 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction