Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 4/CD104 Antibody (439-9B), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143369PE
Description
Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Human, Bovine samples. It is validated for Flow Cytometry.Specifications
Integrin beta 4/CD104 | |
Monoclonal | |
PE | |
P16144 | |
ITGB4 | |
The immunogen for this antibody was CD104. | |
0.1 mL | |
Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
3691 | |
Store at 4C in the dark. |
Flow Cytometry | |
439-9B | |
Flow Cytometry | |
CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Human, Bovine | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction