Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 4/CD104 Antibody (450-9D), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065599PEATT594
Description
Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Human, Ferret, Mustelid samples. It is validated for Flow Cytometry.Specifications
| Integrin beta 4/CD104 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| ITGB4 | |
| Purified alpha6 beta4 integrin from A431 cells | |
| 0.1 mL | |
| Primary | |
| NB100-65599 recognizes the human beta4 integrin (also known as CD104), a 205kD glycoprotein which associates with the alpha6 integrin to form the alpha6/beta4 complex. CD104 is expressed on epithelial cells, Schwann cells and various tumour cell lines. NB100-65599 recognizes an extracellular epitope on the CD104 molecule. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 450-9D | |
| Flow Cytometry | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
| Mouse | |
| Protein G purified | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 3691.0 | |
| Human, Ferret, Mustelid | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction