Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 6 Antibody (268622), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2389MFV610
Description
Integrin beta 6 Monoclonal antibody specifically detects Integrin beta 6 in Mouse samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
Integrin beta 6 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunocytochemistry | |
268622 | |
Western Blot, Immunocytochemistry | |
integrin beta-6, integrin, beta 6 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 6, Gly22-Asn706, Accession # Q9Z0T9 | |
0.1 mL | |
Cancer | |
3694 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction