Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 6 Antibody (437211), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4155Z
Description
Integrin beta 6 Monoclonal antibody specifically detects Integrin beta 6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
Integrin beta 6 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
437211 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
integrin beta-6, integrin, beta 6 | |
Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha V beta 6, Phe31-Val992 (Integrin alpha V) and Gly22-Asn707 (Integrin beta 6), Accession # P18564 | |
0.1 mL | |
Cancer | |
3694 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction