Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 6 Antibody (CSbeta6) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200972 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019468 0.025 mg
NBP229800 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019468 Supplier Novus Biologicals Supplier No. NBP2298000.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin beta 6 Monoclonal specifically detects Integrin beta 6 in Human, Rat, Primate, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready.

Specifications

Antigen Integrin beta 6
Applications CyTOF, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone CSbeta6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, CyTOF-ready
Formulation PBS with No Preservative
Gene Accession No. P18564
Gene Alias integrin beta-6, integrin, beta 6
Gene Symbols ITGB6
Host Species Mouse
Immunogen beta 6 knockout mice have been immunized with recombinant secreted human alpaVbeta6.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3694
Test Specificity Monoclonal antibody MAB2076Z, raised by immunizing beta6 knockout mice with recombinant secreted human alphavbeta6, is useful for flow cytometry and immunoprecipitation in all species tested, including mouse. It also works well for immunohistochemistry of acetone-fixed tissues in several species, including rat, rabbit, monkey, and human. The antibody does not work well for immunohistochemistry of mouse tissue. This antibody does not recognize the aV subunit and does not react with alphaV integrins other than alphavbeta6. Because the antibody is not effective in western Blot, it is unknown whether the epitope recognized by the antibody localizes to the alphavbeta6 heterodimer or to the beta6 subunit only.
Target Species Primate, Rabbit, Rat, Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.