Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 6 Antibody (OTI1D2), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271051PE
Description
Integrin beta 6 Monoclonal antibody specifically detects Integrin beta 6 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Integrin beta 6 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
OTI1D2 | |
Western Blot | |
integrin beta-6, integrin, beta 6 | |
Full length human recombinant protein of human ITGB6(NP_000879) produced in HEK293T cell. | |
0.1 mL | |
Cancer | |
3694 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction