Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 7 Antibody (2G18) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | Integrin beta 7 |
---|---|
Clone | 2G18 |
Concentration | 0.4 mg/mL |
Dilution | Flow Cytometry 1:50-200, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:100-400, Immunohistochemistry-Paraffin 1:100-200 |
Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBT94147100
![]() |
Novus Biologicals
NB11094147100UG |
100 μg |
Each for $560.50
|
|
|||||
NB018055
![]() |
Novus Biologicals
NB110941470.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Integrin beta 7 | |
0.4 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
Mouse | |
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
ITGB7 | |
IgG2 | |
Protein G purified | |
This was selected for its ability to detect mouse CD45 positive leukocytes in IHC. |
2G18 | |
Flow Cytometry 1:50-200, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:100-400, Immunohistochemistry-Paraffin 1:100-200 | |
Monoclonal | |
Purified | |
Signal Transduction | |
A 0.2 um filtered solution in PBS with No Preservative | |
3695 | |
Cell lysates of mouse lymphocytes | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title