Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 7 Antibody (2G18), DyLight 550, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11094147R
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Integrin beta 7 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ITGB7 | |
Cell lysates of mouse lymphocytes | |
0.1 mL | |
Primary | |
This was selected for its ability to detect mouse CD45 positive leukocytes in IHC. | |
Store at 4C in the dark. | |
IgG2 |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2G18 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
Rat | |
Protein A or G purified | |
Signal Transduction | |
3695 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction