Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 7 Antibody (332418), PerCP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB3060C
Description
Integrin beta 7 Monoclonal antibody specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
Integrin beta 7 | |
Monoclonal | |
PerCP | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
332418 | |
Flow Cytometry, CyTOF-ready | |
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 7, Glu20-His722, Accession # P26011 | |
0.1 mL | |
Signal Transduction | |
3695 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction