Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 7 Antibody (473207), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4669APCCY7
Description
Integrin beta 7 Monoclonal antibody specifically detects Integrin beta 7 in Human samples. It is validated for Flow CytometrySpecifications
Integrin beta 7 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
473207 | |
Flow Cytometry | |
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
Chinese hamster ovary cell line CHO-derived recombinant human Integrin beta 7 long isoform, Glu20-His723, Accession # P26010 | |
0.1 mL | |
Signal Transduction | |
3695 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction