Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Integrin beta 7 Chimeric Recombinant Rabbit Monoclonal Antibody (FIB27)
GREENER_CHOICE

Catalog No. PIMA548015
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548015 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548015 Supplier Invitrogen™ Supplier No. MA548015
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: FIB27 binds specifically to both human and mouse integrin beta-7 at epitope region Dl on which is involved in all a4b7-mediated adhesion events. Does not compete with DAKT32 for binding. FIB27 blocks LS722-induced TK1 cell aggregation (mAb LS722 is an activating anti-a4B7 antibody and induces aggregation via an a4B7-dependent pathway).

Integrin beta 7 associates with integrin alpha 4 (CD49d) to form the alpha 4 beta 7 integrin LPAM-1, expressed on intraepithelial lymphocytes. It also associates with alpha E (CD103) to form the alpha E beta 7 integrin HML-1, expressed on T cells adjacent to mucosal epithelium and intraepithelial lymphocytes. Ligands for LPAM-1 include VCAM-1 (CD106), MAdCAM-1 and fibronectin, while the ligand of HML-1 is E-cadherin (CD324). Integrin beta 7 plays an important role in the adhesion of leukocytes to endothelial cells promoting the transmigration of leukocytes to extravascular spaces during the inflammatory response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin beta 7 Chimeric
Applications Flow Cytometry, Inhibition Assays, Immunoprecipitation, Immunocytochemistry
Classification Recombinant Monoclonal
Clone FIB27
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene ITGB7
Gene Accession No. P26010, P26011
Gene Alias Gut homing receptor beta subunit; integrin beta 7; integrin beta 7 subunit; integrin beta-7; Integrin beta-P; integrin subunit beta 7; integrin, beta 7; ITGB7; Ly69; M290 IEL antigen
Gene Symbols ITGB7
Host Species Rabbit
Immunogen FIB27 was prepared by immunizing rats with TK1 cells.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16421, 3695
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.