Learn More
Invitrogen™ Integrin beta 7 Monoclonal Antibody (473207), FITC

Description
In Western blots, no cross-reactivity with recombinant human Integrin beta 1, beta 2, beta 3, beta 4, beta 5, beta 6, beta 8, recombinant mouse Integrin beta 4, or beta 7 is observed.
Specifications
Specifications
| Antigen | Integrin beta 7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 473207 |
| Conjugate | FITC |
| Formulation | Buffered saline solution with BSA and 0.09% sodium azide |
| Gene | ITGB7 |
| Gene Accession No. | P26010 |
| Gene Alias | Gut homing receptor beta subunit; integrin beta 7; integrin beta 7 subunit; integrin beta-7; Integrin beta-P; integrin subunit beta 7; integrin, beta 7; ITGB7; Ly69; M290 IEL antigen |
| Gene Symbols | ITGB7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.