Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 7 Monoclonal Antibody (FIB504), Functional Grade, In Vivo PLATINUM™, Leinco Technologies™
SDP

Catalog No. 502544263
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4263 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4263 Supplier Leinco Technologies Inc Supplier No. B763100MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specificity: Clone FIB504 recognizes an epitope on human/mouse integrin Beta7. Endotoxin Level: <0.5 EU/mg as determined by the LAL method. Purity: ≥98% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco Purified Functional PLATINUM™ antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

Integrin beta 7 Monoclonal antibody specifically detects Integrin beta 7 in Human, Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays, Mass Spectrometry

Specifications

Antigen Integrin beta 7
Applications Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays, Mass Spectrometry
Classification Monoclonal
Clone FIB504
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene ITGB7
Gene Accession No. P26010, P26011
Gene Alias Gut homing receptor beta subunit; integrin beta 7; integrin beta 7 subunit; integrin beta-7; Integrin beta-P; integrin subunit beta 7; integrin, beta 7; ITGB7; Ly69; M290 IEL antigen
Host Species Rat
Immunogen TK1 cells
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16421, 3695
Target Species Human, Mouse
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.