Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Integrin beta 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547440
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547440 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547440 Supplier Invitrogen™ Supplier No. PA547440
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Integrin beta 1, rmIntegrin beta 2, recombinant human Integrin beta 3, and rmIntegrin beta 6 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Integrin beta 7 associates with integrin alpha 4 (CD49d) to form the alpha 4 beta 7 integrin LPAM-1, expressed on intraepithelial lymphocytes. It also associates with alpha E (CD103) to form the alpha E beta 7 integrin HML-1, expressed on T cells adjacent to mucosal epithelium and intraepithelial lymphocytes. Ligands for LPAM-1 include VCAM-1 (CD106), MAdCAM-1 and fibronectin, while the ligand of HML-1 is E-cadherin (CD324). Integrin beta 7 plays an important role in the adhesion of leukocytes to endothelial cells promoting the transmigration of leukocytes to extravascular spaces during the inflammatory response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin beta 7
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ITGB7
Gene Accession No. P26011
Gene Alias Gut homing receptor beta subunit; integrin beta 7; integrin beta 7 subunit; integrin beta-7; Integrin beta-P; integrin subunit beta 7; integrin, beta 7; ITGB7; Ly69; M290 IEL antigen
Gene Symbols ITGB7
Host Species Goat
Immunogen CHO-derived recombinant mouse Integrin beta 7 Glu20-His722.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16421
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.