Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta-like protein 1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Integrin beta-like protein 1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Integrin beta-like protein 1 Polyclonal specifically detects Integrin beta-like protein 1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Integrin beta-like protein 1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
9358 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
integrin, beta-like 1 (with EGF-like repeat domains), OSCPITGBL1, integrin beta-like 1, Osteoblast-specific cysteine-rich protein, Ten integrin EGF-like repeat domain-containing protein, ten integrin EGF-like repeat domains protein, TIEDintegrin beta-like protein 1 | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human Integrin beta-like protein 1 (NP_004782.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title