Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta1 (CD29), Rat anti-Human, Clone: mAb13, Millipore Sigma™

Catalog No. MABT82125UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT82125UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT82125UL Supplier MilliporeSigma Supplier No. MABT82125UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Integrin beta1 (CD29), clone mAb13, Cat. No. MABT821, is a rat monoclonal antibody that detects integrin beta-1 and has been tested for use in Immunocytochemistry, Immunoprecipitation, and Inhibition studies.

Integrin beta-1 (UniProt: P05556; also known as Fibronectin receptor subunit beta, Glycoprotein IIa, GPIIA, VLA-4 subunit beta, CD29) is encoded by the ITGB1 (also known as FNRB, MDF2, MSK12) gene (Gene ID: 3688) in human. Integrin beta-1 is a single-pass type I membrane protein that associates with one of the alpha subunits to form receptor for fibronectin. Integrin beta-1 integrin recognizes the sequence R-G-D in a wide array of ligands. Five isoforms of integrin beta-1 have been reported that are produced by alternative splicing. Integrin beta-1 is synthesized with a signal peptide (aa 1-20) that is subsequently cleaved off. The mature form has an extracellular domain (aa 21-728), a helical domain (aa 729-751) and a cytoplasmic region (aa 752-798). Isoform 1 of integrin beta-1 is widely expressed and other isoforms are generally co-expressed with a more restricted distribution. Isoform 2 is expressed in skin, liver, skeletal muscle, cardiac muscle, placenta, umbilical vein endothelial cells, neuroblastoma cells, lymphoma cells, hepatoma cells and astrocytoma cells. Isoform 3 and isoform 4 are expressed in muscle, kidney, liver, placenta, cervical epithelium, umbilical vein endothelial cells, fibroblast cells, embryonal kidney cells, platelets and several blood cell lines. Isoform 4 is selectively expressed in peripheral T-cells and isoform 5 is expressed specifically in striated muscle (skeletal and cardiac muscle). Isoform 2 is reported to interfere with isoform 1 resulting in a dominant negative effect on cell adhesion and migration (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin beta1 (CD29)
Applications Immunocytochemistry, Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone mAb13
Conjugate Unconjugated
Dilution Inhibition Analysis: A representative lot blocked cell attachment in Inhibition studies (Akiyama, S.K., et. al. (1989). J Cell Biol. 109(2):863-75; Humphries, J.D., et. al. (2005). J Biol Chem. 280(11):10234-43). Immunocytochemistry Analysis: A representative lot detected Integrin beta1 (CD29) in Immunocytochemistry applications (Akiyama, S.K., et. al. (1989). J Cell Biol. 109(2):863-75; Humphries, J.D., et. al. (2005). J Biol Chem. 280(11):10234-43). Immunoprecipitation Analysis: A representative lot detected Integrin beta1 (CD29) in Immunoprecipitation applications (Akiyama, S.K., et. al. (1989). J Cell Biol. 109(2):863-75). Inhibition Analysis: A representative lot blocked  HT-1080 cells adhesion to fibronectin coated plates  (Courtesy of Dr. Kenneth M. Yamada, M.D., Ph.D., NIDCR, National Institutes of Health, Bethesda, MD USA).
Formulation Purified rat monoclonal antibody IgG2a in PBS without preservatives.
Gene Alias Fibronectin receptor subunit beta;Glycoprotein IIa;GPIIA;VLA-4 subunit beta;CD29
Gene Symbols ITGB1;FNRB;MDF2;MSK12
Host Species Rat
Immunogen Fibronectin receptor (alpha V beta 1) purified from human placenta.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone mAb13 specifically detects beta 1 subunit of human integrin. It targets an epitope within 12 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.