Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin αvβ5 Mouse, Brilliant Violet 786, Clone: ALULA, BD Optibuild™

Catalog No. BDB743674 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743674 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743674 Supplier BD Biosciences Supplier No. 743674
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ALULA monoclonal antibody specifically binds to Integrin alpha v beta 5 (αvβ5), which is also known asCD51/β5.Integrin αvβ5 is widely expressed byfibroblasts, endothelial cells, and vascular smooth muscle cells. It is involved in activation-dependent cell migration and cell adhesion to the extracellular matrix. Integrin αvβ5 plays a major role in growth factor-induced angiogenesis in response to VEGF and TGF.The αv Integrins, αvβ5andαvβ3,αre involved in tumor growthand are being targeted in cancer therapies. The ALULA clone specificallyrecognizes αvβ5 integrin and functionally blocks the interaction between αvβ5 and vitronectin.Although raised against mouseαvβ5, the ALULA antibody crossreacts with humanαvβ5.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin αvβ5
Classification Monoclonal
Clone ALULA
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Integrin alpha v beta 5, αVβ5, CD51/β5
Host Species Mouse
Immunogen Mouse fibroblasts expressing αvβ5
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.