Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin αvβ6 Mouse anti-Human, BV510, Clone: 10D5, BD OptiBuild™

Catalog No. BDB751820 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751820 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751820 Supplier BD Biosciences Supplier No. 751820
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin αvβ6 Monoclonal antibody specifically detects Integrin αvβ6 in Human samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin αvβ6
Applications Flow Cytometry
Classification Monoclonal
Clone 10D5
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 510
Description Hu ITGRN ALPVBTA6 BV510 10D5 50 μg
Host Species Mouse
Immunogen 129/C57BL/6 β6 -/- Mice were immunized with Human αvβ6 or Mouse Keratinocytes
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Integrin αvβ6
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a
Reactivity Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.