Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin αX Armenian Hamster anti-Mouse, Unconjugated, Clone: N418, MilliporeSigma™
Armenian Hamster Monoclonal Antibody
$452.71
Specifications
Antigen | Integrin αX |
---|---|
Clone | N418 |
Concentration | Please refer to lot specific datasheet. |
Applications | Immunohistochemistry, Flow Cytometry, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1399Z-I-10
|
MilliporeSigma
MAB1399ZI100UG |
100 μg |
Each of 1 for $452.71
|
|
|||||
Description
Anti-Integrin X, clone N418, azide free, Cat. No. MAB1399Z-I, is an armenian hamster monoclonal antibody that detects Integrin alpha-X and has been tested for use in Immunohistochemistry, Flow Cytometry, and Immunoprecipitation.Specifications
Integrin αX | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
Integrin alpha-X;CD11 antigen-like family member C;Leukocyte adhesion glycoprotein p150;95 alpha chain;Leukocyte adhesion receptor p150;95;CD11c | |
Itgax | |
IgG | |
Protein G purified | |
Clone N418 is a mouse monoclonal antibody that detects integrin alpha-X in murine tissue. | |
129.15 kDa calculated. |
N418 | |
Immunohistochemistry, Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
Armenian Hamster | |
Inflammation & Immunology | |
Purified armenian hamster monoclonal antibody IgG in PBS without preservatives. | |
NP_067309 | |
Mouse spleen dendritic cells. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title