Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Intelectin-1/Omentin Antibody (6C11), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242652B
Description
Intelectin-1/Omentin Monoclonal specifically detects Intelectin-1/Omentin in Human samples. It is validated for Western Blot, ELISA.Specifications
Intelectin-1/Omentin | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
ITLN1 | |
Recombinant human ITLN1 (17-313aa) purified from E. coli | |
100 μL | |
Cardiovascular Biology, metabolism | |
55600 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
6C11 | |
Western Blot, ELISA | |
Endothelial lectin HL-1, FLJ20022, Galactofuranose-binding lectin, hIntL, HL-1, intelectin 1 (galactofuranose binding), intelectin-1, INTLHL1, ITLNITLN-1, LFRIntestinal lactoferrin receptor, omentin | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction