Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Intelectin-1/Omentin Mouse anti-Human, Clone: ITLN1/4063, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Intelectin-1/Omentin |
---|---|
Clone | ITLN1/4063 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
Intelectin-1/Omentin Monoclonal specifically detects Intelectin-1/Omentin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
Intelectin-1/Omentin | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
Endothelial lectin HL-1, FLJ20022, Galactofuranose-binding lectin, hIntL, HL-1, intelectin 1 (galactofuranose binding), intelectin-1, INTLHL1, ITLNITLN-1, LFRIntestinal lactoferrin receptor, omentin | |
Recombinant fragment (around aa16-141) of human Intelectin-1/Omentin protein (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
ITLN1/4063 | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
55600 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title