Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OraSure Technologies InteliSwab™ COVID-19 Rapid Test, OTC

Click to view available options
:
Anterior Nasal Swab
Description
- Self-collected, visual read lateral flow test
- Both symptomatic and asymptomatic use
- Intuitive workflow: just swab, swirl, and see
- Workflow requires < 1 minute of active, hands-on time; 30-minute time to result
- Gentle lower nasal swab
- Overall test performance: Positive Percent Agreement 84%, Negative Percent Agreement 98%
- No precise number of drops required
Specifications
Specifications
CE Marker | No |
Clia Complexity | Waived (OTC) |
Detectable Analytes | SARS-CoV-2 Nucleocapsid Protein Antigen |
Sample Type | Anterior Nasal Swab |
Sensitivity | 84% |
Type | COVID-19 Rapid Antigen Test |
Includes | Security Chip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction