Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OraSure Technologies InteliSwab™ COVID-19 Rapid Test Pro Kit Controls
SDP

Catalog No. 23222114 Shop All OraSure Technologies Products
Change view
Click to view available options
For Use With (Equipment):
InteliSwab™ COVID-19 Rapid Test Pro
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
23-222-114 InteliSwab™ COVID-19 Rapid Test Pro
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-222-114 Supplier OraSure Technologies Supplier No. 10010613
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

External quality control reagents to monitor the performance of InteliSwab™ COVID-19 rapid test pro with direct anterior nasal samples

  • For use only with InteliSwab™ COVID-19 Rapid Test Pro
  • Kit control box contains:
    • Instructions for use (IFU)
    • Specimen collection loops
    • Two vials:
      • One COVID-19 positive control
      • One COVID-19 negative control

Specifications

Content And Storage 36°F to 46°F
Detectable Analytes SARS-CoV-2 Nucleocapsid Protein Antigen
For Use With (Equipment) InteliSwab™ COVID-19 Rapid Test Pro
Type InteliSwab™ Kit Controls
Includes Instructions for use, Specimen collection loops, COVID-19 positive control, COVID-19 negative control

This product has not been FDA cleared or approved, but has been authorized by FDA under an EUA for use by authorized laboratories. This product has been authorized only for the detection of proteins from SARS-CoV-2, not for any other viruses or pathogens. The emergency use of this product is only authorized for the duration of the declaration that circumstances exist justifying the authorization of emergency use of in vitro diagnostics for detection and/or diagnosis of COVID-19 under Section 564(b)(1) of the Federal Food, Drug and Cosmetic Act, 21 U.S.C. 360bbb-3(b)(1), unless the declaration is terminated or authorization is revoked sooner.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.