Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OraSure Technologies InteliSwab™ COVID-19 Rapid Test Pro
SDP

Catalog No. 23222113 Shop All OraSure Technologies Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-222-113 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-222-113 Supplier OraSure Technologies Supplier No. 10010614
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid antigen test provides results in 30 minutes with unique all-in-one test device and nasal swab

  • Self-collected, visual read lateral flow test
  • Both symptomatic and asymptomatic use
  • Intuitive workflow: just swab, swirl and see
  • Workflow requires less than 1 minute of active, hands-on time
    • 30-minute time for results
  • Gentle lower nasal swab
  • Overall test performance:
    • Positive percent agreement 84%
    • Negative percent agreement 98%
  • No pipetting and no precise number of drops required
  • Clinical performance proven with untrained and unproctored lay-users

Specifications

Certifications/Compliance FDA EUA
Content And Storage 35°F to 86°F
Control Sets For purchase separately
CPT Code 87811
CE Marker No
Clia Complexity Waived
Detectable Analytes SARS-CoV-2 Nucleocapsid Protein Antigen
Sample Type Anterior Nasal Swab
Sensitivity 84%
Type COVID-19 Rapid Antigen Test
Packaging Type Kit
Quantity 25 Tests
Specificity 0.98
Test Time 30 min.
Show More Show Less

This product has not been FDA cleared or approved, but has been authorized by FDA under an EUA for use by authorized laboratories. This product has been authorized only for the detection of proteins from SARS-CoV-2, not for any other viruses or pathogens. The emergency use of this product is only authorized for the duration of the declaration that circumstances exist justifying the authorization of emergency use of in vitro diagnostics for detection and/or diagnosis of COVID-19 under Section 564(b)(1) of the Federal Food, Drug and Cosmetic Act, 21 U.S.C. 360bbb-3(b)(1), unless the declaration is terminated or authorization is revoked sooner.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.