Learn More
Honeywell IntelliDoX Docking Station
Honeywell IntelliDoX Docking Station is a bump test and calibration station for gas detectors that demonstrates quick and efficient performance.
Supplier:  Honeywell DXMCLP
 View more versions of this product
Catalog No. 19-807-577
Only null left
Add to Cart
Description
Description
Honeywell IntelliDoX Docking Station is a bump test and calibration station for gas detectors that demonstrates quick and efficient performance.
  • High-tech way to combine smart docking modules and Fleet Manager II software to provide automated instrument management to drive productivity, reliability and efficiency.
  • Enables bump test tracking, email notices and easy recordkeeping for improved compliance
  • Performs a quick bump test for increased uptime and no wasted calibration gas
  • Enables the ability to change alarm set points, set a noncompliance indicator when a bump test is due, and adjust other parameters for maximum flexibility
  • IntelliDoX module comes with factory-installed nest
  • Power cord, calibration gas tubing and exhaust tubing are packaged separately
  • One Enabler Kit is required for each stand-alone IntelliDoX module
  • 2-year warranty
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Certicate of Compliance; CSA/UL 61010-1 (3rd Ed); EC Declaration of Conformity; EMC Directive & Low Voltage Directive
BW MicroClip Series
13.8 cm
1.91 kg
13.8 x 36.2 x 10.9 cm
Gas Detection Monitor Test Stations
36.2 cm
10.9 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.